Methyl 3-(methylthio)propionate, 98%
Methyl 3-(methylthio)propionate, 98%
Methyl 3-(methylthio)propionate, 98%
Thermo Scientific Chemicals

Methyl 3-(methylthio)propionate, 98%

CAS: 13532-18-8 | C5H10O2S | 134.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10649.36
also known as A10649-36
Price (USD)
206.65
Online Exclusive
229.00
Save 22.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
206.65
Online Exclusive
229.00
Save 22.35 (10%)
Each
Chemical Identifiers
CAS13532-18-8
IUPAC Namemethyl 3-(methylsulfanyl)propanoate
Molecular FormulaC5H10O2S
InChI KeyDMMJVMYCBULSIS-UHFFFAOYSA-N
SMILESCOC(=O)CCSC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Refractive Index1.4635-1.4675 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only