2,5-Difluorobenzoyl chloride, 97+%
2,5-Difluorobenzoyl chloride, 97+%
2,5-Difluorobenzoyl chloride, 97+%
Thermo Scientific Chemicals

2,5-Difluorobenzoyl chloride, 97+%

CAS: 35730-09-7 | C7H3ClF2O | 176.547 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A10651.06
also known as A10651-06
Price (USD)
43.65
Special offer
Online exclusive
Ends: 30-Jun-2026
50.60
Save 6.95 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
43.65
Special offer
Online exclusive
Ends: 30-Jun-2026
50.60
Save 6.95 (14%)
Each
Chemical Identifiers
CAS35730-09-7
IUPAC Name2,5-difluorobenzoyl chloride
Molecular FormulaC7H3ClF2O
InChI KeyRLRUKKDFNWXXRT-UHFFFAOYSA-N
SMILESFC1=CC=C(F)C(=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.0%
Identification (FTIR)Conforms
Refractive Index1.5115-1.5165 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only