2,5-Diphenyloxazole, 99%
2,5-Diphenyloxazole, 99%
2,5-Diphenyloxazole, 99%
Thermo Scientific Chemicals

2,5-Diphenyloxazole, 99%

CAS: 92-71-7 | C15H11NO | 221.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10654.36
also known as A10654-36
Price (USD)
312.65
Special offer
Online exclusive
Ends: 30-Jun-2026
368.00
Save 55.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
312.65
Special offer
Online exclusive
Ends: 30-Jun-2026
368.00
Save 55.35 (15%)
Each
Chemical Identifiers
CAS92-71-7
IUPAC Name2,5-diphenyl-1,3-oxazole
Molecular FormulaC15H11NO
InChI KeyCNRNYORZJGVOSY-UHFFFAOYSA-N
SMILESO1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (HPLC)≥98.5%
Identification (FTIR)Conforms
Melting Point (clear melt)69.0-75.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only