1-Bromopinacolone, 97+%
1-Bromopinacolone, 97+%
1-Bromopinacolone, 97+%
Thermo Scientific Chemicals

1-Bromopinacolone, 97+%

CAS: 5469-26-1 | C6H11BrO | 179.057 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A10655.09
also known as A10655-09
Price (USD)
73.30
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
73.30
Each
Chemical Identifiers
CAS5469-26-1
IUPAC Name1-bromo-3,3-dimethylbutan-2-one
Molecular FormulaC6H11BrO
InChI KeySAIRZMWXVJEBMO-UHFFFAOYSA-N
SMILESCC(C)(C)C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4630-1.4680 @ 20?C
FormLiquid
Assay (GC)≥97.0%
1-Bromopinacolone is used as an iIntermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopinacolone is used as an iIntermediate for triazole compounds ( a five-membered ring structure compound contains three nitrogens and two carbons in the ring) which have biological activities such as antiviral, antibacterial, antifungal and antituberculous.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases, heat and light.
RUO – Research Use Only
Saul G.Cohen; Deborah L.Lieberman; Fariza B.Hasan; Jonathan B.Cohen. 1-Bromopinacolone, an active site-directed covalent inhibitor for acetylcholinesterase. The journal of Biological Chemistry. 1982, 257, (23),14087-14092.