Ethyl decanoate, 99%
Ethyl decanoate, 99%
Ethyl decanoate, 99%
Thermo Scientific Chemicals

Ethyl decanoate, 99%

CAS: 110-38-3 | C12H24O2 | 200.322 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A10657.0E
also known as A10657-0E
Price (USD)
387.65
Special offer
Online exclusive
Ends: 30-Jun-2026
456.00
Save 68.35 (15%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
387.65
Special offer
Online exclusive
Ends: 30-Jun-2026
456.00
Save 68.35 (15%)
Each
Chemical Identifiers
CAS110-38-3
IUPAC Nameethyl decanoate
Molecular FormulaC12H24O2
InChI KeyRGXWDWUGBIJHDO-UHFFFAOYSA-N
SMILESCCCCCCCCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4240-1.4270 @ 20?C
Assay (GC)≥98.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only