2-Bromopropiophenone, 96%
2-Bromopropiophenone, 96%
2-Bromopropiophenone, 96%
Thermo Scientific Chemicals

2-Bromopropiophenone, 96%

CAS: 2114-00-3 | C9H9BrO | 213.074 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10661.36
also known as A10661-36
Price (USD)
719.65
Special offer
Online exclusive
Ends: 30-Jun-2026
846.00
Save 126.35 (15%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
719.65
Special offer
Online exclusive
Ends: 30-Jun-2026
846.00
Save 126.35 (15%)
Each
Chemical Identifiers
CAS2114-00-3
IUPAC Name2-bromo-1-phenylpropan-1-one
Molecular FormulaC9H9BrO
InChI KeyWPDWOCRJBPXJFM-UHFFFAOYNA-N
SMILESCC(Br)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥95.0%
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow to green to orange
Refractive Index1.5690-1.5750 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only