2-Benzylpyridine, 98+%
2-Benzylpyridine, 98+%
2-Benzylpyridine, 98+%
Thermo Scientific Chemicals

2-Benzylpyridine, 98+%

CAS: 101-82-6 | C12H11N | 169.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A10662.30
also known as A10662-30
Price (USD)
86.65
Online Exclusive
96.00
Save 9.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
86.65
Online Exclusive
96.00
Save 9.35 (10%)
Each
Chemical Identifiers
CAS101-82-6
IUPAC Name2-benzylpyridine
Molecular FormulaC12H11N
InChI KeyPCFUWBOSXMKGIP-UHFFFAOYSA-N
SMILESC(C1=CC=CC=C1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.5775-1.5815 @ 20?C
Appearance (Color)Clear colorless to pale yellow
2-Benzylpyridine is used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Benzylpyridine is used in organic synthesis.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Katsuma. Hiraki.; Yoshio. Fuchita.; Keizo. Takechi. Preparation and characterization of novel six-membered cyclopalladated complexes of 2-benzylpyridine. Inorg. Chem. 1981, 20, (12), 4316-4320.
  2. William Henderson.; Brian K Nicholson.; Suzanne J Faville.; Daming Fan.; John D Ranford. Gold(III) thiosalicylate complexes containing cycloaurated 2-arylpyridine, 2-anilinopyridine and 2-benzylpyridine ligands. Journal of Organometallic Chemistry. 2001, 631 (1-2), 41-46.