2-Bromobenzonitrile, 99%
2-Bromobenzonitrile, 99%
2-Bromobenzonitrile, 99%
Thermo Scientific Chemicals

2-Bromobenzonitrile, 99%

CAS: 2042-37-7 | C7H4BrN | 182.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10679.06
also known as A10679-06
Price (USD)
61.65
Online Exclusive
68.10
Save 6.45 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
61.65
Online Exclusive
68.10
Save 6.45 (9%)
Each
Chemical Identifiers
CAS2042-37-7
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)49-56?C
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only