4-Bromo-2,6-dimethylaniline, 98%
4-Bromo-2,6-dimethylaniline, 98%
4-Bromo-2,6-dimethylaniline, 98%
4-Bromo-2,6-dimethylaniline, 98%
Thermo Scientific Chemicals

4-Bromo-2,6-dimethylaniline, 98%

CAS: 24596-19-8 | C8H10BrN | 200.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
50 g
250 g
Catalog number A10712.14
also known as A10712-14
Price (USD)
86.20
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
86.20
Each
Add to cart
Chemical Identifiers
CAS24596-19-8
IUPAC Name4-bromo-2,6-dimethylaniline
Molecular FormulaC8H10BrN
InChI KeyQGLAYJCJLHNIGJ-UHFFFAOYSA-N
SMILESCC1=CC(Br)=CC(C)=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown to dark brown or grey
Melting Point (clear melt)46.0-53.0°C
FormCrystals or powder or crystalline powder or lumps
Assay (GC)≥97.5%
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only