4-Iodobenzoic acid, 97%
4-Iodobenzoic acid, 97%
4-Iodobenzoic acid, 97%
Thermo Scientific Chemicals

4-Iodobenzoic acid, 97%

CAS: 619-58-9 | C7H5IO2 | 248.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A10730.30
also known as A10730-30
Price (USD)
538.65
Special offer
Online exclusive
Ends: 30-Jun-2026
634.00
Save 95.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
538.65
Special offer
Online exclusive
Ends: 30-Jun-2026
634.00
Save 95.35 (15%)
Each
Chemical Identifiers
CAS619-58-9
IUPAC Name4-iodobenzoic acid
Molecular FormulaC7H5IO2
InChI KeyGHICCUXQJBDNRN-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)267.0-276.0?C
Appearance (Color)White to pale cream to pale orange to pale brown
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
4-Iodobenzoic acid was used in the synthesis of [hydroxy(4-carboxyphenyl)iodonium]ion in situ that helps in the cleavage of a variety of alkenes. It is is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzoic acid was used in the synthesis of [hydroxy(4-carboxyphenyl)iodonium]ion in situ that helps in the cleavage of a variety of alkenes. It is is used as anti-infective, contraceptive agent and x-ray contrast medium for diagnostic radiology.

Solubility
Partly miscible with water. Soluble in alcohol & ether.

Notes
It is sensitive to light. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Fadi Homsi; Kazushi Hosoi; Kyoko Nozaki; Tamejiro Hiyama. Solid phase cross-coupling reaction of aryl(halo)silanes with 4-iodobenzoic acid. Journal of Organometallic Chemistry. 2001, 624, (1-2)208-216
  2. Cara L.Nygren; Chick C.Wilson; John F.C.Turner. On the Solid State Structure of 4-Iodobenzoic Acid. J. Phys. Chem. A. 2005, 109, (11),2586-2593