3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt, 98%
3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt, 98%
3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt, 98%
Thermo Scientific Chemicals

3,5-Dichloro-2-hydroxybenzenesulfonic acid sodium salt, 98%

CAS: 54970-72-8 | C6H3Cl2NaO4S | 265.036 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A10732.09
also known as A10732-09
Price (USD)
54.65
Online Exclusive
60.50
Save 5.85 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
54.65
Online Exclusive
60.50
Save 5.85 (10%)
Each
Chemical Identifiers
CAS54970-72-8
IUPAC Namesodium 3,5-dichloro-2-hydroxybenzene-1-sulfonic acid
Molecular FormulaC6H4Cl2NaO4S
InChI KeyNMWCVZCSJHJYFW-UHFFFAOYSA-N
SMILES[Na+].OC1=C(Cl)C=C(Cl)C=C1S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only