1-Cyclohexene-1-carboxylic acid, 97%
1-Cyclohexene-1-carboxylic acid, 97%
1-Cyclohexene-1-carboxylic acid, 97%
Thermo Scientific Chemicals

1-Cyclohexene-1-carboxylic acid, 97%

CAS: 636-82-8 | C7H10O2 | 126.155 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A10741.03
also known as A10741-03
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.70
Save 7.05 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.70
Save 7.05 (15%)
Each
Chemical Identifiers
CAS636-82-8
IUPAC Namecyclohex-1-ene-1-carboxylic acid
Molecular FormulaC7H10O2
InChI KeyNMEZJSDUZQOPFE-UHFFFAOYSA-N
SMILESOC(=O)C1=CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystalline powder or fused solid or clear liquid as melt
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
Melting Point (clear melt)27.0-39.0°C
View more
1-Cyclohexene-1-carboxylic acid is an intermediate in benzoic acid anaerobic decomposition by methanogenic consortium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Cyclohexene-1-carboxylic acid is an intermediate in benzoic acid anaerobic decomposition by methanogenic consortium.

Solubility
Soluble in water (0.7 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
RUO – Research Use Only

General References:

  1. Gholamreza Bahrami.; Bahareh Mohammadi.; Amir Kiani. Determination of oseltamivir carboxylic acid in human serum by solid phase extraction and high performance liquid chromatography with UV detection. Journal of Chromatography B. 1986, 864, (1-2), 38-42.
  2. K. C. Nicolaou.; A. J. Roecker.; Jeffrey A. Pfefferkorn.; Gou-Qiang Cao. A Novel Strategy for the Solid-Phase Synthesis of Substituted Indolines. J. Am. Chem. Soc. 2000, 122 (12), 2966-2967.