2-Chloro-4-iodoaniline, 98%
2-Chloro-4-iodoaniline, 98%
2-Chloro-4-iodoaniline, 98%
2-Chloro-4-iodoaniline, 98%
Thermo Scientific Chemicals

2-Chloro-4-iodoaniline, 98%

CAS: 42016-93-3 | C6H5ClIN | 253.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10742.06
also known as A10742-06
Price (USD)
50.65
Online Exclusive
56.50
Save 5.85 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
50.65
Online Exclusive
56.50
Save 5.85 (10%)
Each
Add to cart
Chemical Identifiers
CAS42016-93-3
IUPAC Name2-chloro-4-iodoaniline
Molecular FormulaC6H5ClIN
InChI KeyMYDAOWXYGPEPJT-UHFFFAOYSA-N
SMILESNC1=CC=C(I)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown to dark brown to gray
Melting Point (clear melt)67.0-74.0°C
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only