This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Solution TestClear to slightly turbid colorless to yellow (10% in Ethanol, non-U.S. specification)
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥97.5%
Melting Point (clear melt)51.5-57.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.