2-Chlorobenzotrifluoride, 99%
2-Chlorobenzotrifluoride, 99%
2-Chlorobenzotrifluoride, 99%
Thermo Scientific Chemicals

2-Chlorobenzotrifluoride, 99%

CAS: 88-16-4 | C7H4ClF3 | 180.554 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A10753.36
also known as A10753-36
Price (USD)
57.65
Online Exclusive
63.40
Save 5.75 (9%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
57.65
Online Exclusive
63.40
Save 5.75 (9%)
Each
Chemical Identifiers
CAS88-16-4
IUPAC Name1-chloro-2-(trifluoromethyl)benzene
Molecular FormulaC7H4ClF3
InChI KeyDGRVQOKCSKDWIH-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4545-1.4575 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only