Manganese(IV) oxide, 97%
Manganese(IV) oxide, 97%
Manganese(IV) oxide, 97%
Thermo Scientific™

Manganese(IV) oxide, 97%

CAS: 1313-13-9 | MnO2 | 86.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
10 kg
Catalog number A10765.A9
also known as A10765-A9
Price (USD)
504.65
Online Exclusive
561.00
Save 56.35 (10%)
Each
Quantity:
10 kg
Request bulk or custom format
Price (USD)
504.65
Online Exclusive
561.00
Save 56.35 (10%)
Each
Chemical Identifiers
CAS1313-13-9
IUPAC Namedioxomanganese
Molecular FormulaMnO2
InChI KeyNUJOXMJBOLGQSY-UHFFFAOYSA-N
SMILESO=[Mn]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to dark grey to black
FormPowder
Complexometric Titration≥96.0 to ≤104.0%
Manganese(IV) oxide is used as a pigment and as a reagent inorganic synthesis especially for the oxidation of allylic alcohols. It is also used for dry -cell batteries such as the alkaline battery and the zinc-carbon battery. It is involved in the preparation of other manganese compounds. It is a precursor to ferromanganese and related alloys, which is useful in the steel industry. It catalyses the decomposition of hydrogen peroxide into oxygen and water. Further, it is used to prepare manganese(II) sulfate, manganese(II) chloride and potassium permanganate. In addition to this, it is used in ceramics and glass manufacturing as an inorganic pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(IV) oxide is used as a pigment and as a reagent inorganic synthesis especially for the oxidation of allylic alcohols. It is also used for dry -cell batteries such as the alkaline battery and the zinc-carbon battery. It is involved in the preparation of other manganese compounds. It is a precursor to ferromanganese and related alloys, which is useful in the steel industry. It catalyses the decomposition of hydrogen peroxide into oxygen and water. Further, it is used to prepare manganese(II) sulfate, manganese(II) chloride and potassium permanganate. In addition to this, it is used in ceramics and glass manufacturing as an inorganic pigment.

Solubility
Insoluble in water.

Notes
Incompatible with strong acids, strong reducing agents and organic materials.
RUO – Research Use Only

General References:

  1. Zhang, C.; Liang, P.; Jiang, Y.; Huang, X. Enhanced power generation of microbial fuel cell using manganese dioxide-coated anode in flow-through mode. J. Power Sources 2015, 273, 580-583.
  2. Huang, M.; Zhao, X. L.; Li, F.; Zhang, L. L.; Zhang, Y. X. Facile synthesis of ultrathin manganese dioxide nanosheets arrays on nickel foam as advanced binder-free supercapacitor electrodes. J. Power Sources 2015, 277, 36-43.