Pyridine-3,4-dicarboxylic anhydride, 97%
Pyridine-3,4-dicarboxylic anhydride, 97%
Pyridine-3,4-dicarboxylic anhydride, 97%
Pyridine-3,4-dicarboxylic anhydride, 97%
Thermo Scientific Chemicals

Pyridine-3,4-dicarboxylic anhydride, 97%

CAS: 4664-08-8 | C7H3NO3 | 149.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A10771.03
also known as A10771-03
Price (USD)
35.65
Online Exclusive
39.80
Save 4.15 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
35.65
Online Exclusive
39.80
Save 4.15 (10%)
Each
Add to cart
Chemical Identifiers
CAS4664-08-8
IUPAC Name1H,3H-furo[3,4-c]pyridine-1,3-dione
Molecular FormulaC7H3NO3
InChI KeyKFKMGUPDWTWQFM-UHFFFAOYSA-N
SMILESO=C1OC(=O)C2=CN=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused solid
Acid-base back titration≥96.0 to ≤104.0%
Appearance (Color)White to cream to pale brown
Pyridine-3,4-dicarboxylic anhydride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-3,4-dicarboxylic anhydride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Decomposes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents. It is sensitive to moisture.
RUO – Research Use Only

General References:

  1. A. Paul Krapcho.; Mary E. Petry.; Zelleka Getahun.; John J. LandiJr.; John Stallman.; Johanna F. Polsenberg.; Cynthia E. Gallagher.; Martin J. Maresch, Miles P. Hacker. 6,9-Bis[(aminoalkyl)amino]benzo[g]isoquinoline-5,10-diones. A Novel Class of Chromophore-Modified Antitumor Anthracene-9,10-diones: Synthesis and Antitumor Evaluations. J. Med. Chem. 1994, 37, (6), 828-837.
  2. Kenji Niiyama.; Toshiaki Mase.; Hirobumi Takahashi.; Akira Naya.; Kasumi Katsuki.; Toshio Nagase.; Satoshi Ito.; Takashi Hayama.; Akihiro Hisaka.; Satoshi Ozaki.; Masaki Ihara.; Mitsuo Yano.; Takahiro Fukuroda.; Kazuhito Noguchi.; Masaru Nishikibe.; Kiyofumi Ishikawa. 6-Carboxy-5,7-diarylcyclopenteno[1,2-b]pyridine derivatives: A novel class of endothelin receptor antagonists. Bioorganic & Medicinal Chemistry. 2002, 10 (8), 2461-2470.