4,5-Dimethylthiazole, 98%
4,5-Dimethylthiazole, 98%
4,5-Dimethylthiazole, 98%
4,5-Dimethylthiazole, 98%
Thermo Scientific Chemicals

4,5-Dimethylthiazole, 98%

CAS: 3581-91-7 | C5H7NS | 113.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10785.22
also known as A10785-22
Price (USD)
333.65
Special offer
Online exclusive
Ends: 30-Jun-2026
392.00
Save 58.35 (15%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
333.65
Special offer
Online exclusive
Ends: 30-Jun-2026
392.00
Save 58.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS3581-91-7
IUPAC Name4,5-dimethyl-1,3-thiazole
Molecular FormulaC5H7NS
InChI KeyUWSONZCNXUSTKW-UHFFFAOYSA-N
SMILESCC1=C(C)N=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to yellow
FormLiquid
Refractive Index1.5195-1.5235 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only