2-Phenoxyethanol, 94%
2-Phenoxyethanol, 94%
2-Phenoxyethanol, 94%
2-Phenoxyethanol, 94%
Thermo Scientific Chemicals

2-Phenoxyethanol, 94%

CAS: 122-99-6 | C8H10O2 | 138.166 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A10786.30
also known as A10786-30
Price (USD)
26.65
Online Exclusive
29.90
Save 3.25 (11%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
26.65
Online Exclusive
29.90
Save 3.25 (11%)
Each
Add to cart
Chemical Identifiers
CAS122-99-6
IUPAC Name2-phenoxyethan-1-ol
Molecular FormulaC8H10O2
InChI KeyQCDWFXQBSFUVSP-UHFFFAOYSA-N
SMILESOCCOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid or viscous liquid
Assay (GC)≥92.0%
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to white to pale yellow
Refractive Index1.5340-1.5410 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Phenoxyethanol is used as a solvent for cellulose acetate, inks and resins. It is also used as a fixative for perfumes, insect repellents and topical antiseptics. It is utilized as a surface active agent. It acts as an anesthetic in the aquaculture of some fish. Further, it is used as a preservative and cosmetic raw material. In addition to this, it is used in preparation of active pharmaceutical ingredients and dyes.

Solubility
Miscible with water, ethanol, propanol and propylene glycol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Cao, G.; Sheng, D.; Xu, W.; Wang, X. Structural and dyeing properties of aramid treated with 2-phenoxyethanol. Color. Technol. 2015, 131 (5), 384-388.
  2. Toni, C.; Martos-Sitcha, J. A.; Baldisserotto, B.; Heinzmann, B. M.; de Lima Silva, L.; Martínez-Rodríguez, G.; Mancera, J. M. Sedative effect of 2-phenoxyethanol and essential oil of Lippia alba on stress response in gilthead sea bream (Sparus aurata). Res. Vet. Sci. 2015, 103, 20-27.