NEW - Major Software Update Available!—Cell Cycle Analysis can be performed using the Tali® Image-Based Cytometer
NEW - Tali® Cell Cycle Kit Launched! Read More!
Product Image
Invitrogen™

Tali™ Apoptosis Kit - Annexin V Alexa Fluor™ 488 & Propidium Iodide

The Tali™ Apoptosis Kit enables identification of apoptotic cells and discrimination of apoptotic from necrotic and live cells in aRead more
Have Questions?
Catalog NumberQuantity
A107881 kit
Catalog number A10788
Price (USD)
542.00
Each
Quantity:
1 kit
Price (USD)
542.00
Each
The Tali™ Apoptosis Kit enables identification of apoptotic cells and discrimination of apoptotic from necrotic and live cells in a population. The kit stains apoptotic cells with green Annexin V – Alexa Fluor™ 488, stains necrotic cells with both red propidium iodide and green Annexin V – Alexa Fluor™ 488, and does not stain live cells.

The Tali™Apoptosis Kit contains:

Annexin V – Alexa Fluor™ 488 conjugate

• Annexin Binding solution
• Propidium Iodide (PI) solution

Apoptosis is a carefully regulated process of cell death that occurs as a normal part of development. Inappropriately regulated apoptosis is implicated in disease states, such as Alzheimer’s disease and cancer. Apoptosis is distinguished from necrosis by characteristic morphological and biochemical changes, including compaction and fragmentation of the nuclear chromatin, shrinkage of the cytoplasm, and loss of membrane asymmetry.

Easily Discriminate Apoptotic from Necrotic Cells with Two-color Staining
In normal live cells, phosphatidyl serine (PS) is located on the cytoplasmic surface of the cell membrane. In apoptotic cells, however, PS is translocated from the inner to the outer surface of the plasma membrane, exposing it to the extracellular environment. The human anticoagulant Annexin V displays a high affinity for PS. Annexin V labeled with a fluorophore such as Alexa Fluor™ 488 can be used to identify apoptotic cells by binding to the exposed PS.

The Tali™ Apoptosis Kit includes a solution of propidium Iodide (PI). PI is a cell-impermeant fluorogenic DNA-binding dye used for years to identify necrotic cells. PI is impermeant to live cells, but easily enters dead cells where it binds to nucleic acids and becomes fluorescent.

After staining a cell population using the Tali™ Apoptosis Assay Kit with Annexin V-Alexa Fluor™ 488 and Propidium Iodide, apoptotic cells display green fluorescence, dead cells display red and green fluorescence and live cells show little or no fluorescence.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Tali™ Image-Based Cytometer
Product LineTali
Product TypeApoptosis Kit
Quantity1 kit
Shipping ConditionWet Ice
ConjugateAlexa Fluor 488
FormatVial
Unit SizeEach
Contents & Storage
Tali™ - Annexin V Alexa Fluor™: 488: Store 2 – 6°C
Tali™ - Propidium Iodide: Store 2 – 6°C
Tali™ - Annexin V Binding Buffer: Store 2 – 6°C

Frequently asked questions (FAQs)

What are the fluorescence excitation/emission maxima for the dyes contained in the Tali Apoptosis Kit - Annexin V Alexa Fluor 488 & Propidium Iodide?

Annexin V Alexa Fluor 488: 488/499 nm
Propidium Iodide: 535/617 nm, bound to DNA

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (2)

Citations & References
Abstract
Gain of 20q11.21 in human embryonic stem cells improves cell survival by increased expression of Bcl-xL.
Authors:Nguyen HT, Geens M, Mertzanidou A, Jacobs K, Heirman C, Breckpot K, Spits C,
Journal:Mol Hum Reprod
PubMed ID:24217388
Gain of 20q11.21 is a chromosomal abnormality that is recurrently found in human pluripotent stem cells and cancers, strongly suggesting that this mutation confers a proliferative or survival advantage to these cells. In this work we studied three human embryonic stem cell (hESC) lines that acquired a gain of 20q11.21 ... More
STAT3 and MCL-1 associate to cause a mesenchymal epithelial transition.
Authors:Renjini AP, Titus S, Narayan P, Murali M, Kumar Jha R, Laloraya M,
Journal:
PubMed ID:24481815
'Embryo implantation is effected by a myriad of signaling cascades acting on the embryo-endometrium axis. Here we show, by using MALDI TOF analysis, far-western analysis and colocalization and co-transfection studies, that STAT3 and MCL-1 are interacting partners during embryo implantation. We show in vitro that the interaction between the two ... More