Thermo Scientific Chemicals

3-Aminobenzamide, 98%, Thermo Scientific Chemicals

Catalog number: A10793.14
25 g, Each
Thermo Scientific Chemicals

3-Aminobenzamide, 98%, Thermo Scientific Chemicals

Catalog number: A10793.14
25 g, Each
Quantity
Catalog number: A10793.14
also known as A10793-14
Price (USD)
Price: 215.00
Online price: 185.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3544-24-9
IUPAC Name
3-aminobenzamide
Molecular Formula
C7H8N2O
InChI Key
GSCPDZHWVNUUFI-UHFFFAOYSA-N
SMILES
NC(=O)C1=CC=CC(N)=C1
Melting Point (clear melt)
110.0-116.0?C
Appearance (Color)
Pale cream to cream to pale brown
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (GC)
≥97.5%

Description

3-Aminobenzamide is used as an endogenous poly-ADP-ribosyltransferase inhibitor. Through its effects on PARPs, 3-amino benzamide causes telomere shortening and stimulates angiogenesis. It displays neuroprotective, anti-necrotic effects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminobenzamide is used as an endogenous poly-ADP-ribosyltransferase inhibitor. Through its effects on PARPs, 3-amino benzamide causes telomere shortening and stimulates angiogenesis. It displays neuroprotective, anti-necrotic effects.

Solubility
Soluble in dimethylformamide (∼30 mg/ml), dimethyl sulfoxide (∼30 mg/ml), phosphate buffered saline (pH7.2) (∼2 mg/ml), water (25 mg/ml), and ethanol (25 mg/ml).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text