4-Fluoro-3-nitrobenzoic acid, 98%
4-Fluoro-3-nitrobenzoic acid, 98%
4-Fluoro-3-nitrobenzoic acid, 98%
Thermo Scientific Chemicals

4-Fluoro-3-nitrobenzoic acid, 98%

CAS: 453-71-4 | C7H4FNO4 | 185.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10800.06
also known as A10800-06
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
48.70
Save 7.05 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
48.70
Save 7.05 (14%)
Each
Chemical Identifiers
CAS453-71-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow to pale gray
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)120.0-126.0?C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only