4-Bromo-2-fluorophenol, 97+%
4-Bromo-2-fluorophenol, 97+%
4-Bromo-2-fluorophenol, 97+%
Thermo Scientific Chemicals

4-Bromo-2-fluorophenol, 97+%

CAS: 2105-94-4 | C6H4BrFO | 191.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10804.06
also known as A10804-06
Price (USD)
23.20
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
23.20
Each
Chemical Identifiers
CAS2105-94-4
IUPAC Name4-bromo-2-fluorophenol
Molecular FormulaC6H4BrFO
InChI KeyRYVOZMPTISNBDB-UHFFFAOYSA-N
SMILESOC1=CC=C(Br)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.0%
Refractive Index1.5640-1.5690 @ 20?C
Appearance (Color)Clear colorless to yellow to pale brown
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only