N-Boc-L-tyrosine, 98+%, Thermo Scientific Chemicals
N-Boc-L-tyrosine, 98+%, Thermo Scientific Chemicals
N-Boc-L-tyrosine, 98+%, Thermo Scientific Chemicals
N-Boc-L-tyrosine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-L-tyrosine, 98+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
A10810.03
also known as A10810-03
1g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10810.03
also known as A10810-03
Price (USD)
-
Quantity:
1g
Request bulk or custom format
Chemical Identifiers
CAS3978-80-1
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
Molecular FormulaC14H19NO5
InChI KeyCNBUSIJNWNXLQQ-LDGXTIHJNA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSpecification differs for U.S. and non-U.S. material where indicated
Optical Rotation34.0-38.0° (C=1, Dioxane, U.S. specification)
Optical Rotation37.0 ± 0.1° (C=1 in dioxane, non-U.S. specification)
Appearance (Color)White to pale cream
FormPowder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only