4-Phenylphenol, 97%
4-Phenylphenol, 97%
4-Phenylphenol, 97%
Thermo Scientific Chemicals

4-Phenylphenol, 97%

CAS: 92-69-3 | C12H10O | 170.211 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A10817.22
also known as A10817-22
Price (USD)
63.80
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
63.80
Each
Chemical Identifiers
CAS92-69-3
IUPAC Name[1,1'-biphenyl]-4-ol
Molecular FormulaC12H10O
InChI KeyYXVFYQXJAXKLAK-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale brown or pale pink
Melting Point (clear melt)163.0-170.0°C
Assay (GC)≥96.0%
FormPowder and/or flakes
Fluorescence and phosphorescence quantum yields and fluorescence and phosphorescence lifetimes were obtained for 4-phenylphenol adsorbed on filter paper with either NaCl, NaBr, or NaI at 296 and 93 K. The solid-surface fluorescence and phosphorescence quantum yield values and phosphorescence lifetime values were obtained for p-aminobenzoic acid (PABA) and 4-phenylphenol adsorbed on α-cyclodextrin/NaCl mixtures. 4-Phenylphenol is used as an antioxidant and is a potential EDC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorescence and phosphorescence quantum yields and fluorescence and phosphorescence lifetimes were obtained for 4-phenylphenol adsorbed on filter paper with either NaCl, NaBr, or NaI at 296 and 93 K. The solid-surface fluorescence and phosphorescence quantum yield values and phosphorescence lifetime values were obtained for p-aminobenzoic acid (PABA) and 4-phenylphenol adsorbed on α-cyclodextrin/NaCl mixtures. 4-Phenylphenol is used as an antioxidant and is a potential EDC.

Solubility
Soluble in water (0.7 mg/ml at 20°C), ether, most common organic solvents, methanol, and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Peng Xu,; Jayant Kumar,; Lynne Samuelsonm,; Ashok L. Cholli. Monitoring the Enzymatic Polymerization of 4-Phenylphenol by Matrix-Assisted Laser Desorption Ionization Time-of-Flight Mass Spectrometry: A Novel Approach. Biomacromolecules. 2002, 3(5), 889-893.
  2. S.M. Ramasamy,; R.J. Hurtubise.Temperature effects on the solid-matrix luminescence properties of 4-phenylphenol adsorbed on filter paper. Talanta. 1989, 36(1-2),315-320.