1-Bromopentane, 99%
1-Bromopentane, 99%
1-Bromopentane, 99%
Thermo Scientific Chemicals

1-Bromopentane, 99%

CAS: 110-53-2 | C5H11Br | 151.047 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10821.30
also known as A10821-30
Price (USD)
49.30
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
49.30
Each
Chemical Identifiers
CAS110-53-2
IUPAC Name1-bromopentane
Molecular FormulaC5H11Br
InChI KeyYZWKKMVJZFACSU-UHFFFAOYSA-N
SMILESCCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4420-1.4460 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥98.5%
1-Bromopentane is used in a multitude of organic synthesis. It is used in iron-catalyzed alkylations of aromatic Grignard reagents via cross coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromopentane is used in a multitude of organic synthesis. It is used in iron-catalyzed alkylations of aromatic Grignard reagents via cross coupling.

Solubility
Insoluble in water and soluble in alcohol. Miscible in diethyl ether.

Notes
Store away from oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Keiichi Ohno.; Hiroshi Yoshida.; Hiroatsu Matsuura. Conformational dependence of the isolated C-D stretching and CHD rocking vibrations of selectively monodeuterated alkyl chains as proved by conformational polymorphism of 1-bromopentane. Spectrochimica Acta Part A: Molecular and Biomolecular Spectroscopy. 1996, 52 (11), 1377-1389.
  2. John P. Idouxa.; Ronald Wysockia.; Sherre Younga.; Jeffrey Turcota.; Charlene Ohlmana.; Russell Leonard. Polymer-Supported Multi-Site Phase Transfer Catalysts. Synthetic Communications: An International Journal for Rapid Communication of Synthetic Organic Chemistry. 1983, 13 (2), 139-144.