2-Amino-4-methylbenzothiazole, 98%
2-Amino-4-methylbenzothiazole, 98%
2-Amino-4-methylbenzothiazole, 98%
Thermo Scientific Chemicals

2-Amino-4-methylbenzothiazole, 98%

CAS: 1477-42-5 | C8H8N2S | 164.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10822.22
also known as A10822-22
Price (USD)
146.65
Special offer
Online exclusive
Ends: 30-Jun-2026
172.00
Save 25.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
146.65
Special offer
Online exclusive
Ends: 30-Jun-2026
172.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS1477-42-5
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to pale cream
Melting Point (clear melt)135.0-142.0?C
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only