2-Chlorobenzoic acid, 98+%
2-Chlorobenzoic acid, 98+%
2-Chlorobenzoic acid, 98+%
2-Chlorobenzoic acid, 98+%
Thermo Scientific Chemicals

2-Chlorobenzoic acid, 98+%

CAS: 118-91-2 | C7H5ClO2 | 156.565 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A10832.30
also known as A10832-30
Price (USD)
26.65
Special offer
Online exclusive
Ends: 30-Jun-2026
31.70
Save 5.05 (16%)
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)
26.65
Special offer
Online exclusive
Ends: 30-Jun-2026
31.70
Save 5.05 (16%)
Each
Add to cart
Chemical Identifiers
CAS118-91-2
IUPAC Name2-chlorobenzoic acid
Molecular FormulaC7H5ClO2
InChI KeyIKCLCGXPQILATA-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥98.0%
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Melting Point (clear melt)137.0-143.0?C
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only