Primary Rat Cortex Neurons, 1 x 106 cells - FAQs
Primary Rat Cortex Neurons, 1 x 106 cells - FAQs
View additional product information for Primary Rat Cortex Neurons - FAQs (A1084002, A1084001)
6 product FAQs found
What is the difference between Primary Rat Cortex Neurons, Sprague Dawley and Primary Rat Cortex Neurons?
These neurons differ in the rat strain from which they were isolated: Sprague Dawley vs. Fisher 344. These neurons don't expand anymore. They just grow neurites. The manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0001574-PrimaryRatCortex-HippocampusNeurons-UG.pdf) has instructions for culturing the cells. You can use Neurobasal Plus/B-27 Plus/GlutaMAX to culture them, or just Neurobasal/B-27/GlutaMAX. However, the former combination provides much superior performance than the latter and can maintain neurons longer.
I'm currently culturing neurons. Which media system do you recommend using in conjunction with the Poly-D-Lysine matrix?
We recommend using the B-27 Plus Neuronal Culture System when culturing neurons on Poly-D-Lysine. The B-27 Plus Neuronal Culture System is a complete media system that includes the B-27 Plus Supplement and the Neurobasal Plus Medium.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What cell types will Poly-D-Lysine support as an attachment matrix?
The Poly-D-Lysine matrix has successfully supported the attachment of primary rodent neurons, iPSC-derived neurons and PC12 cells.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can primary rat neurons be cryopreserved again after they are cultured?
Primary rat neurons cannot be cryopreserved again because they will not revive.
Can primary rat neurons proliferate?
No, primary rat neurons do not proliferate.
Why does the manual for primary rat neurons have the instruction "Do not expose neurons to air at any time"?
Neurons tend to lift off from the complete medium change, which exposes cells to air. By changing half of the medium, the disturbance can be minimized.