3-Chloro-4-fluorothiophenol, 97%
3-Chloro-4-fluorothiophenol, 97%
3-Chloro-4-fluorothiophenol, 97%
3-Chloro-4-fluorothiophenol, 97%
Thermo Scientific Chemicals

3-Chloro-4-fluorothiophenol, 97%

CAS: 60811-23-6 | C6H3ClFS | 161.60 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A10856.06
also known as A10856-06
Price (USD)
303.65
Special offer
Online exclusive
Ends: 30-Jun-2026
357.00
Save 53.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
303.65
Special offer
Online exclusive
Ends: 30-Jun-2026
357.00
Save 53.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS60811-23-6
IUPAC Name(3-chloro-4-fluorophenyl)sulfanide
Molecular FormulaC6H3ClFS
InChI KeySFSHSIFYTWIGSF-UHFFFAOYSA-M
SMILESFC1=CC=C([S-])C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
Refractive Index1.5705-1.5755 @ 20?C
FormLiquid
Used as pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharamaceutical intermediate.

Solubility
Sparingly Soluble (0.014 g/L) (25°C).

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only

General References:

  1. Giancarlo Angelini,; Gaetano Lilla,; Maurizio Speranza. Gas-phase heteroaromatic substitution. 3. Electrophilic methylation of furan and thiophene by CH3XCH3+ (X = fluoro, chloro) ions. J. Am. Chem. Soc., 1982, 104(25) ,7091-7098.
  2. J. R. Lacher,; Ralph E. Scruby, J. D. Park.Diamagnetic Susceptibilities of Some Chloro and Fluoro Derivatives of Methane and Ethylene. J. Am. Chem. Soc., 1949, 71(5), 1797-1799.