1-Bromo-3-fluorobenzene, 99%
1-Bromo-3-fluorobenzene, 99%
1-Bromo-3-fluorobenzene, 99%
Thermo Scientific Chemicals

1-Bromo-3-fluorobenzene, 99%

CAS: 1073-06-9 | C6H4BrF | 175 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
100 g
250 g
Catalog number A10858.18
also known as A10858-18
Price (USD)
70.65
Special offer
Online exclusive
Ends: 30-Jun-2026
82.60
Save 11.95 (14%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
70.65
Special offer
Online exclusive
Ends: 30-Jun-2026
82.60
Save 11.95 (14%)
Each
Chemical Identifiers
CAS1073-06-9
IUPAC Name1-bromo-3-fluorobenzene
Molecular FormulaC6H4BrF
InChI KeyQDFKKJYEIFBEFC-UHFFFAOYSA-N
SMILESFC1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Assay (GC)≥98.5%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5260-1.5290 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only