2,6-Dichlorobenzamide, 98%
2,6-Dichlorobenzamide, 98%
2,6-Dichlorobenzamide, 98%
Thermo Scientific Chemicals

2,6-Dichlorobenzamide, 98%

CAS: 2008-58-4 | C7H5Cl2NO | 190.023 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10865.22
also known as A10865-22
Price (USD)
175.65
Online Exclusive
195.00
Save 19.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
175.65
Online Exclusive
195.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS2008-58-4
IUPAC Name2,6-dichlorobenzamide
Molecular FormulaC7H5Cl2NO
InChI KeyJHSPCUHPSIUQRB-UHFFFAOYSA-N
SMILESNC(=O)C1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Melting Point (clear melt)195.0-204.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only