2,3,5-Triphenyl-2H-tetrazolium chloride, 98%
2,3,5-Triphenyl-2H-tetrazolium chloride, 98%
2,3,5-Triphenyl-2H-tetrazolium chloride, 98%
Thermo Scientific Chemicals

2,3,5-Triphenyl-2H-tetrazolium chloride, 98%

2,3,5-Triphenyl-2H-tetrazolium chloride, 98%, C19H15ClN4, CAS Number-298-96-4, triphenyltetrazolium chloride, tetrazolium chloride, urocheck, tetrazolium red, tetrazolium chloride, uroscreen, red tetrazolium, vitastain, 2,3,5-triphenyltetrazolium chloride, 2,3,5-triphenyl-2h-tetrazolium chloride | CAS: 298-96-4 | C19H15ClN4 | 334.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A10870.30
Price (USD)
772.65
Online Exclusive
858.00
Save 85.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
772.65
Online Exclusive
858.00
Save 85.35 (10%)
Each
Chemical Identifiers
CAS298-96-4
IUPAC Nametriphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular FormulaC19H15ClN4
InChI KeyPKDBCJSWQUOKDO-UHFFFAOYSA-M
SMILES[Cl-].C1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or cream
FormCrystals or crystalline powder or powder
Identification (FTIR)Conforms
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)≤1.0%
2,3,5-Triphenyl-2H-tetrazolium chloride is used as a redox indicator. It is also used in biochemical experiments, especially to indicate cellular respiration. It is employed in autopsy pathology to assist identification of post-mortem myocardial infarctions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5-Triphenyl-2H-tetrazolium chloride is used as a redox indicator. It is also used in biochemical experiments, especially to indicate cellular respiration. It is employed in autopsy pathology to assist identification of post-mortem myocardial infarctions.

Solubility
Soluble in water, ethanol and acetone. Insoluble in ether.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Reagent for detection of reducing sugars.
  2. Oxidizing agent, reduced to the colored formazan, capable of introducing a -hydroxyl substituent into steroidal enones: J. Chem. Soc., Perkin 1, 2687 (1988).
  3. McLaughlin, M. R.; Balaa, M. F. Enhanced contrast of bacteriophage plaques in Salmonella with ferric ammonium citrate and sodium thiosulfate (FACST) and tetrazolium red (TZR). J. Microbiol. Methods 2006, 65 (2), 318-323.
  4. Li, X.; Deng, S.; Fu, H. Synergism between red tetrazolium and uracil on the corrosion of cold rolled steel in H2SO4 solution. Corros. Sci. 2009, 51 (6), 1344-1355.