3-Chlorobenzoic acid, 99%
3-Chlorobenzoic acid, 99%
3-Chlorobenzoic acid, 99%
Thermo Scientific Chemicals

3-Chlorobenzoic acid, 99%

CAS: 535-80-8 | C7H5ClO2 | 156.565 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10876.14
also known as A10876-14
Price (USD)
28.65
Online Exclusive
32.10
Save 3.45 (11%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
28.65
Online Exclusive
32.10
Save 3.45 (11%)
Each
Chemical Identifiers
CAS535-80-8
IUPAC Name3-chlorobenzoic acid
Molecular FormulaC7H5ClO2
InChI KeyLULAYUGMBFYYEX-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
Melting Point (clear melt)153.0-159.0°C
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%

A Pseudomonas species which utilized 3-chlorobenzoic acid as a carbon source converted this compound to 3-hydroxybenzoic and 2, 5-dihydroxybenzoic acids. A methanogenic consortium able to use 3-chlorobenzoic acid as its sole energy and carbon source was enriched from anaerobic sewage sludge. A Pseudomonas putida strain 87 capable of assimilating 3-chlorobenzoic acid as a sole source of carbon and energy (3Cba+) was isolated. coupling of 3-chlorobenzoic acid with phenylboronic acid proceeds at room temperature using 0.5 % Pd and at 100°C using 0.1 % Pd to provide the coupled product in 97 % yeild.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A Pseudomonas species which utilized 3-chlorobenzoic acid as a carbon source converted this compound to 3-hydroxybenzoic and 2, 5-dihydroxybenzoic acids. A methanogenic consortium able to use 3-chlorobenzoic acid as its sole energy and carbon source was enriched from anaerobic sewage sludge. A Pseudomonas putida strain 87 capable of assimilating 3-chlorobenzoic acid as a sole source of carbon and energy (3Cba+) was isolated. coupling of 3-chlorobenzoic acid with phenylboronic acid proceeds at room temperature using 0.5 % Pd and at 100°C using 0.1 % Pd to provide the coupled product in 97 % yeild.

Solubility
Slioghtly soluble in water(0.45 g/L ).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. N. Walker, D. Harris. Metabolism of 3-chlorobenzoic acid by Azotobacter species. Soil Biology and Biochemistry. 1970, 2(1), 27-32.
  2. Pascale Clément,; Dietmar H. Pieper,; Bernardo González. Molecular characterization of a deletion/duplication rearrangement in tfd genes from Ralstonia eutropha JMP134(pJP4) that improves growth on 3-chlorobenzoic acid but abolishes growth on 2,4-dichlorophenoxyacetic acid. Microbiology,. 2001, 147(8), 2141-2148,o.
  3. The unprotected acid undergoes lithiation at the 2-position with 2.2 moles of s-BuLi in the presence of TMEDA; subsequent reaction with dibromotetrachloroethane provides access to 2-bromo-3-chlorobenzoic acid: Tetrahedron Lett., 36, 881 (1995); Bull. Soc. Chim. Fr., 133, 133 (1996).