2-Amino-6-chlorobenzoic acid, 97+%
2-Amino-6-chlorobenzoic acid, 97+%
2-Amino-6-chlorobenzoic acid, 97+%
Thermo Scientific Chemicals

2-Amino-6-chlorobenzoic acid, 97+%

CAS: 2148-56-3 | C7H6ClNO2 | 171.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
Catalog number A10890.06
also known as A10890-06
Price (USD)
66.65
Online Exclusive
74.20
Save 7.55 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
74.20
Save 7.55 (10%)
Each
Chemical Identifiers
CAS2148-56-3
IUPAC Name2-amino-6-chlorobenzoic acid
Molecular FormulaC7H6ClNO2
InChI KeySZCPTRGBOVXVCA-UHFFFAOYSA-N
SMILESNC1=CC=CC(Cl)=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormPowder
Assay (HPLC)≥97.5%
Appearance (Color)Cream to pale brown
2-Amino-6-chlorobenzoic acid is used as intermediate for medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-chlorobenzoic acid is used as intermediate for medicine.

Solubility
Very soluble in water. Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

General References:

  1. Miriam E. Rogers.; Bruce A. Averill. Symmetrically trisubstituted triptycenes. J. Org. Chem. 1986, 51, (17), 3308-3314.
  2. Yi Jin.; Hui-Yuan Li.; Li-Ping Lin.; Jinzhi Tan.; Jian Ding.; Xiaomin Luo.; Ya-Qiu Long. Synthesis and antitumor evaluation of novel 5-substituted-4-hydroxy-8-nitroquinazolines as EGFR signaling-targeted inhibitors. Bioorganic & Medicinal Chemistry. 2005, 13, (19), 5613-5622.