2'-Aminoacetophenone, 98%
2'-Aminoacetophenone, 98%
2'-Aminoacetophenone, 98%
Thermo Scientific Chemicals

2'-Aminoacetophenone, 98%

CAS: 551-93-9 | C8H9NO | 135.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
Catalog number A10895.18
also known as A10895-18
Price (USD)
211.65
Online Exclusive
235.00
Save 23.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
211.65
Online Exclusive
235.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS551-93-9
IUPAC Name1-(2-aminophenyl)ethan-1-one
Molecular FormulaC8H9NO
InChI KeyGTDQGKWDWVUKTI-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow or orange to dark brown
FormClear liquid or low melting solid
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Refractive Index1.6125-1.6180 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only