2-Ethylhexane-1,3-diol, 97%
2-Ethylhexane-1,3-diol, 97%
2-Ethylhexane-1,3-diol, 97%
Thermo Scientific Chemicals

2-Ethylhexane-1,3-diol, 97%

CAS: 94-96-2 | C8H18O2 | 146.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10,000 mL
500 mL
2500 mL
Catalog number A10898.AP
also known as A10898-AP
Price (USD)
55.65
Online Exclusive
61.50
Save 5.85 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
55.65
Online Exclusive
61.50
Save 5.85 (10%)
Each
Chemical Identifiers
CAS94-96-2
IUPAC Name2-ethylhexane-1,3-diol
Molecular FormulaC8H18O2
InChI KeyRWLALWYNXFYRGW-UHFFFAOYNA-N
SMILESCCCC(O)C(CC)CO
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4485-1.4535 @ 20?C
Assay (Silylated GC)≥96.0%
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
2-Ethylhexane-1,3-diol is used in organic synthesis and can be used to produce polyester resin, polyester plasticizers, polyurethane resin. It is also effective vermifuge to mosquitoes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexane-1,3-diol is used in organic synthesis and can be used to produce polyester resin, polyester plasticizers, polyurethane resin. It is also effective vermifuge to mosquitoes.

Solubility
Soluble in water (42g/L at 20°C). Soluble in alcohol and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is hygroscopic in nature.
RUO – Research Use Only

General References:

  1. Georg Troll.; Alfred Sauerer. Determination of trace amounts of boron in geological samples with carminic acid after extraction with 2-ethylhexane-1,3-diol. Analyst. 1985, 110, (3), 283-286.
  2. Keith R. Betty.; Gerald T. Day. Spectrophotometric determination of boron in nuclear-grade uranium oxides with curcumin after extraction with 2-ethylhexane-1,3-diol. Analyst. 1986, 111, (4), 455-457.