Methyl 2-(methylthio)benzoate, 98%
Methyl 2-(methylthio)benzoate, 98%
Methyl 2-(methylthio)benzoate, 98%
Methyl 2-(methylthio)benzoate, 98%
Thermo Scientific Chemicals

Methyl 2-(methylthio)benzoate, 98%

CAS: 3704-28-7 | C9H10O2S | 182.237 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A10905.03
also known as A10905-03
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
48.70
Save 7.05 (14%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 30-Jun-2026
48.70
Save 7.05 (14%)
Each
Add to cart
Chemical Identifiers
CAS3704-28-7
IUPAC Namemethyl 2-(methylsulfanyl)benzoate
Molecular FormulaC9H10O2S
InChI KeyCPQDZXPLQXZJGF-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1SC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormPowder
Appearance (Color)Pale cream
Melting Point (clear melt)63.0-69.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only