Diethylenetriaminepentaacetic acid, 98+%
Diethylenetriaminepentaacetic acid, 98+%
Diethylenetriaminepentaacetic acid, 98+%
Thermo Scientific Chemicals

Diethylenetriaminepentaacetic acid, 98+%

CAS: 67-43-6 | C14H23N3O10 | 393.349 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
5000 g
Catalog number A10926.0B
also known as A10926-0B
Price (USD)
113.00
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
113.00
Each
Chemical Identifiers
CAS67-43-6
IUPAC Name2-[bis({2-[bis(carboxymethyl)amino]ethyl})amino]acetic acid
Molecular FormulaC14H23N3O10
InChI KeyQPCDCPDFJACHGM-UHFFFAOYSA-N
SMILESOC(=O)CN(CCN(CC(O)=O)CC(O)=O)CCN(CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (unspecified)≥98.0%
FormPowder
Identification (FTIR)Conforms
Diethylenetriaminepentaacetic acid is used as chelating agent and forms complexes with metal ions. It finds application as a magnetic resonance imaging (MRI) contrasting agent, since it improves the image by forming a complex with a gadolinium ion. It can also be used for treatment of radioactive materials such as plutonium, americium and other actinides. It is an active component of manganese and zinc fertilizers used in agrochemical industry. It acts as a color inhibitor and is used in the production of acrylon. It is also used in soaps as a water softener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylenetriaminepentaacetic acid is used as chelating agent and forms complexes with metal ions. It finds application as a magnetic resonance imaging (MRI) contrasting agent, since it improves the image by forming a complex with a gadolinium ion. It can also be used for treatment of radioactive materials such as plutonium, americium and other actinides. It is an active component of manganese and zinc fertilizers used in agrochemical industry. It acts as a color inhibitor and is used in the production of acrylon. It is also used in soaps as a water softener.

Solubility
Soluble in water and alkali.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Chelating agent.
  2. Fitzsimmons, J.; Jay, M. Preparative-scale separation of ethyl esters of diethylenetriaminepentaacetic acid by flash chromatography. J. Radioanal. Nucl. Chem. 2015, 305 (1), 329-336.
  3. Bhatt, R.; Sreedhar, B.; Padmaja, P. Adsorption of chromium from aqueous solutions using crosslinked chitosan-diethylenetriaminepentaacetic acid. Int. J. Biol. Macromol. 2015, 74, 458-466.