Azomethine-H monosodium salt hydrate
Azomethine-H monosodium salt hydrate
Azomethine-H monosodium salt hydrate
Thermo Scientific Chemicals

Azomethine-H monosodium salt hydrate

CAS: 206752-32-1 | C17H12NNaO8S2 | 445.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A10927.14
Price (USD)
209.65
Online Exclusive
233.00
Save 23.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
209.65
Online Exclusive
233.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS206752-32-1
IUPAC Namesodium 4-hydroxy-5-{[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}-7-sulfonaphthalene-2-sulfonate
Molecular FormulaC17H12NNaO8S2
InChI KeyGRYBZNLNHVSHPY-UHFFFAOYSA-M
SMILES[Na+].OC1=C2C(NC=C3C=CC=CC3=O)=CC(=CC2=CC(=C1)S([O-])(=O)=O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Assay (ex Sulphated Ash)> 85.0% (with reference to dried material)
Water Content (Karl Fischer Titration)≤ 8.0%
Formpowder
Azomethine-H monosodium salt hydrate is a reagent used for the colorimetric determination of boron in soil, plants, composts, manure, water and nutrient solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azomethine-H monosodium salt hydrate is a reagent used for the colorimetric determination of boron in soil, plants, composts, manure, water and nutrient solutions.

Solubility
Soluble in water, ethanol and acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.
RUO – Research Use Only

General References:

  1. Xin Rui Wang.; Jun Lu.; Dongpeng Yan.; Min Wei.; David G. Evans.; Xue Duan. A photochromic thin film based on salicylideneaniline derivatives intercalated layered double hydroxide. Chemical Physics Letters. 2010, 493, (4-6), 333-339.
  2. Reagent for the colorimetric determination of boron: Analyst, 94, 1135 (1969); Anal. Lett., 8, 559 (1975).