Diethyl acetone-1,3-dicarboxylate, 96%
Diethyl acetone-1,3-dicarboxylate, 96%
Diethyl acetone-1,3-dicarboxylate, 96%
Thermo Scientific Chemicals

Diethyl acetone-1,3-dicarboxylate, 96%

CAS: 105-50-0 | C9H14O5 | 202.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A10933.30
also known as A10933-30
Price (USD)
154.65
Special offer
Online exclusive
Ends: 28-Dec-2026
182.00
Save 27.35
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
154.65
Special offer
Online exclusive
Ends: 28-Dec-2026
182.00
Save 27.35
Each
Chemical Identifiers
CAS105-50-0
IUPAC Name1,5-diethyl 3-oxopentanedioate
Molecular FormulaC9H14O5
InChI KeyZSANYRMTSBBUCA-UHFFFAOYSA-N
SMILESCCOC(=O)CC(=O)CC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4375-1.4435 @ 20?C
Assay (GC)≥95.0%
Identification (FTIR)Conforms
Synthesis of 1-substituted 4-ethoxycarbonyl-5- (ethoxycarbonylmethyl) pyrazoles Diethyl acetone-1, 3-dicarboxylate reacts with N, N-dimethylforma- mide dimethyl acetal (DMFDMA) in ethanol at room temperature. Unexpected product dichotomy is produced in the Biginelli-like condensation of 2-hydroxybenzaldehyde with urea or thiourea and dimethyl or diethyl acetone-1,3-dicarboxylate, respectively, as active methylene components. Ethyl 2-amino-4-(2-ethoxy-2-oxoethyl)thiazole-5-carboxylate (2a), prepared from diethylacetone-1,3-dicarboxylate, sulfuryl chloride and thiourea. Synthesis of diethyl 2,2-diethyl-3,5-dioxopimelate by the reaction of ethyl 3-chloro-3-oxo-2,2-dimethylpropionate with diethyl acetone-1,3-dicarboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Synthesis of 1-substituted 4-ethoxycarbonyl-5- (ethoxycarbonylmethyl) pyrazoles Diethyl acetone-1, 3-dicarboxylate reacts with N, N-dimethylforma- mide dimethyl acetal (DMFDMA) in ethanol at room temperature. Unexpected product dichotomy is produced in the Biginelli-like condensation of 2-hydroxybenzaldehyde with urea or thiourea and dimethyl or diethyl acetone-1,3-dicarboxylate, respectively, as active methylene components. Ethyl 2-amino-4-(2-ethoxy-2-oxoethyl)thiazole-5-carboxylate (2a), prepared from diethylacetone-1,3-dicarboxylate, sulfuryl chloride and thiourea. Synthesis of diethyl 2,2-diethyl-3,5-dioxopimelate by the reaction of ethyl 3-chloro-3-oxo-2,2-dimethylpropionate with diethyl acetone-1,3-dicarboxylate.

Solubility
Insoluble in water and soluble in alcohol, ester, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. T. A. Bryson,; T. M. Dolak. Dimethyl 2,3-Pentadienedioate. Organic Syntheses.
  2. Stefanie Reima,; Van Thi Hong Nguyen,; Uwe Albrechta,; Peter Langer. Synthesis of 3,5-dioxoalkanoates, 3,5-dioxopimelates and 2,4-dioxoadipates by acylation of 1,3-bis-silyl enol ethers. Tetrahedron Letters. 2005, 46 (48),8423-8425.
  3. Valuable synthetic building block, compare Dimethyl acetone-1,3-dicarboxyl ate, A14969. Used in synthesis of isoquinolin-2-ones: Heterocycles, 33, 515 (1992).