1-Butyl isothiocyanate, 98+%
1-Butyl isothiocyanate, 98+%
1-Butyl isothiocyanate, 98+%
Thermo Scientific Chemicals

1-Butyl isothiocyanate, 98+%

CAS: 592-82-5 | C5H9NS | 115.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A10935.14
also known as A10935-14
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS592-82-5
IUPAC Name1-isothiocyanatobutane
Molecular FormulaC5H9NS
InChI KeyLIMQQADUEULBSO-UHFFFAOYSA-N
SMILESCCCCN=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.4985-1.5025 @ 20?C
1-Butyl isothiocyanate participates in the synthesis and characterization of Poly(N-Propargylthiourea). Participates in the Synthesis of 2-Aminobenzothiazoles via Copper (I)-Catalyzed Cross-Coupling with Part-Per-Million Catalyst Loadings. Thiols reactions with 1-Butyl isothiocyanate in aquatic toxicity data in the TETRATOX assay and reactivity data in an abiotic thiol assay. utyl isothiocyanate has been used as capping reagent to immobilize the functionalized derivative of Oregon Green 514 molecular switch on a preformed amino self assembled monlayer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butyl isothiocyanate participates in the synthesis and characterization of Poly(N-Propargylthiourea). Participates in the Synthesis of 2-Aminobenzothiazoles via Copper (I)-Catalyzed Cross-Coupling with Part-Per-Million Catalyst Loadings. Thiols reactions with 1-Butyl isothiocyanate in aquatic toxicity data in the TETRATOX assay and reactivity data in an abiotic thiol assay. utyl isothiocyanate has been used as capping reagent to immobilize the functionalized derivative of Oregon Green 514 molecular switch on a preformed amino self assembled monlayer.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.
RUO – Research Use Only

General References:

  1. T. Wayne Schultz,; Jason W. Yarbrough,; Tyler B. Pilkington. Aquatic toxicity and abiotic thiol reactivity of aliphatic isothiocyanates: Effects of alkyl-size and -shape . Environmental Toxicology and Pharmacology. 2007, 232 (1), 10-17.
  2. P Mela et. al. Monolayer-functionalized microfluidics devices for optical sensing of acidity.. Lab On a Chip. 2005, 5 (2), 163-170.