2,5-Dibromopyridine, 97%
2,5-Dibromopyridine, 97%
2,5-Dibromopyridine, 97%
Thermo Scientific Chemicals

2,5-Dibromopyridine, 97%

CAS: 624-28-2 | C5H3Br2N | 236.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A10937.09
Price (USD)
48.65
Special offer
Online exclusive
Ends: 30-Jun-2026
56.60
Save 7.95 (14%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
48.65
Special offer
Online exclusive
Ends: 30-Jun-2026
56.60
Save 7.95 (14%)
Each
Chemical Identifiers
CAS624-28-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point92.0-97.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only