2-Methyl-6-nitroquinoline, 98%
2-Methyl-6-nitroquinoline, 98%
2-Methyl-6-nitroquinoline, 98%
Thermo Scientific Chemicals

2-Methyl-6-nitroquinoline, 98%

CAS: 613-30-9 | C10H8N2O2 | 188.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A10945.03
also known as A10945-03
Price (USD)
72.65
Online Exclusive
80.30
Save 7.65 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
80.30
Save 7.65 (10%)
Each
Chemical Identifiers
CAS613-30-9
IUPAC Name2-methyl-6-nitroquinoline
Molecular FormulaC10H8N2O2
InChI KeyDXDPHHQJZWWAEH-UHFFFAOYSA-N
SMILESCC1=CC=C2C=C(C=CC2=N1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow or cream to brown to dark brown/green
FormCrystals or crystalline powder or powder
Melting Point162-167°C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only