6-Hydroxy-2,4-dimethylpyrimidine, 99%
6-Hydroxy-2,4-dimethylpyrimidine, 99%
6-Hydroxy-2,4-dimethylpyrimidine, 99%
Thermo Scientific Chemicals

6-Hydroxy-2,4-dimethylpyrimidine, 99%

CAS: 6622-92-0 | C6H8N2O | 124.143 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A10960.06
also known as A10960-06
Price (USD)
25.65
Special offer
Online exclusive
Ends: 30-Jun-2026
29.50
Save 3.85 (13%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
25.65
Special offer
Online exclusive
Ends: 30-Jun-2026
29.50
Save 3.85 (13%)
Each
Chemical Identifiers
CAS6622-92-0
IUPAC Name2,6-dimethyl-1,4-dihydropyrimidin-4-one
Molecular FormulaC6H8N2O
InChI KeyUQFHLJKWYIJISA-UHFFFAOYSA-N
SMILESCC1=CC(=O)N=C(C)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting Point (clear melt)195-202?C
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
FormCrystalline powder or crystals or needles
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only