(R)-(+)-1-Phenylethylamine, 98%
(R)-(+)-1-Phenylethylamine, 98%
(R)-(+)-1-Phenylethylamine, 98%
Thermo Scientific Chemicals

(R)-(+)-1-Phenylethylamine, 98%

CAS: 3886-69-9 | C8H11N | 121.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A10967.14
also known as A10967-14
Price (USD)
57.65
Online Exclusive
63.40
Save 5.75 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
57.65
Online Exclusive
63.40
Save 5.75 (9%)
Each
Chemical Identifiers
CAS3886-69-9
IUPAC Name1-phenylethan-1-amine
Molecular FormulaC8H11N
InChI KeyRQEUFEKYXDPUSK-UHFFFAOYNA-N
SMILESCC(N)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Optical Rotation29.0 ± 1.5? (C=10 in ethanol)
FormLiquid
View more
(R)-(+)-1-Phenylethylamine is a chiral amine used for the determination of the enantiomeric purity of acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1-Phenylethylamine is a chiral amine used for the determination of the enantiomeric purity of acids.

Solubility
Soluble in water (40 g/L).

Notes
Air sensitive. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
RUO – Research Use Only

General References:

  1. Louis Provencher; Hla Wynn; J.Bryan Jones; Andrzej R.Krawczyk. Enzymes in organic synthesis 51. Probing the dimensions of the large hydrophobic pocket of the active site of pig liver esterase. Tetrahedron: Asymmetry. 1993, 4,(9), 2025-2040.
  2. T.Hudlicky; Gene Gillman; Catherine Andersen. Homochiral amine synthesis by baker's yeast resolution of a β-keto amide: 1-Phenylethylamine. Tetrahedron: Asymmetry. 1992, 3,(2), 281-286.