4-Hydroxyphenylacetonitrile, 97%
4-Hydroxyphenylacetonitrile, 97%
4-Hydroxyphenylacetonitrile, 97%
Thermo Scientific Chemicals

4-Hydroxyphenylacetonitrile, 97%

CAS: 14191-95-8 | C8H7NO | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A10971.03
also known as A10971-03
Price (USD)
31.65
Online Exclusive
34.90
Save 3.25 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
31.65
Online Exclusive
34.90
Save 3.25 (9%)
Each
Chemical Identifiers
CAS14191-95-8
IUPAC Name2-(4-hydroxyphenyl)acetonitrile
Molecular FormulaC8H7NO
InChI KeyAYKYOOPFBCOXSL-UHFFFAOYSA-N
SMILESOC1=CC=C(CC#N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to dark cream or yellow
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥96.0%
Melting Point (clear melt)64.0-74.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only