Cyclopentene, 97%
Cyclopentene, 97%
Cyclopentene, 97%
Thermo Scientific Chemicals

Cyclopentene, 97%

CAS: 142-29-0 | C5H8 | 68.119 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number A11027.AE
also known as A11027-AE
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
56.00
Save 8.35 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
56.00
Save 8.35 (15%)
Each
Chemical Identifiers
CAS142-29-0
IUPAC Namecyclopentene
Molecular FormulaC5H8
InChI KeyLPIQUOYDBNQMRZ-UHFFFAOYSA-N
SMILESC1CC=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4195-1.4245 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Cyclopentene is used in the production of cyclopentene oxide, ketamine, cyclopentyl phenol (disinfectant), aldehyde and cyclopentene glycol. It acts as a monomer and is utilized for the synthesis of plastics and in various chemical syntheses. It is also used in the investigation of [2+2] cycloaddition of diamond (001) surfaces with alkene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentene is used in the production of cyclopentene oxide, ketamine, cyclopentyl phenol (disinfectant), aldehyde and cyclopentene glycol. It acts as a monomer and is utilized for the synthesis of plastics and in various chemical syntheses. It is also used in the investigation of [2+2] cycloaddition of diamond (001) surfaces with alkene.

Solubility
Miscible with ethanol, ether, benzene and organic solvents. Immiscible with water.

Notes
Store in cool place. Air sensitive. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Narayana, G. V.; Xu, G.; Wang, D.; Frey, W.; Buchmeiser, M. R. Access to Ultra-High-Molecular Weight Poly(ethylene) and Activity Boost in the Presence of Cyclopentene With Group 4 Bis-Amido Complexes. ChemPlusChem 2014, 79 (1), 151-162.
  2. Nacereddine, A. K.; Layeb, H.; Chafaa, F.; Yahia, W.; Djerourou, A.; Domingo, L. R. A DFT study of the role of the Lewis acid catalysts in the [3 + 2] cycloaddition reaction of the electrophilic nitrone isomer of methyl glyoxylate oxime with nucleophilic cyclopentene. RSC Adv. 2015, 5 (79), 64098-64105.