Diethylenetriamine, 99%
Diethylenetriamine, 99%
Diethylenetriamine, 99%
Thermo Scientific Chemicals

Diethylenetriamine, 99%

CAS: 111-40-0 | C4H13N3 | 103.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2500 mL
Catalog number A11033.0F
also known as A11033-0F
Price (USD)
119.00
Each
Quantity:
2500 mL
Request bulk or custom format
Price (USD)
119.00
Each
Chemical Identifiers
CAS111-40-0
IUPAC Namebis(2-aminoethyl)amine
Molecular FormulaC4H13N3
InChI KeyRPNUMPOLZDHAAY-UHFFFAOYSA-N
SMILESNCCNCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow or pale pink or pale brown
Assay (GC)≥98.5% (UK-sourced material)
Assay from Supplier's CofA≥98.5% (US-sourced Material)
CommentMaterial sourced in UK and US
Identification (FTIR)Conforms (UK-sourced material)
View more
Diethylenetriamine is used as a solvent for plastics, dyes and in chemical synthesis. It serves as a curing agent for epoxy-resins. As a tridentate ligand, it form complexes such as Co(dien)(NO2)3 in coordination chemistry. It is also used as a fuel additive. It reacts with fatty acid resulting in the formation of amidoamines, which are used as corrosion inhibitors in the petroleum industry. It is associated with unsymmetrical dimethylhydrazine to prepare hydyne, which is a propellant for liquid-fuel rockets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylenetriamine is used as a solvent for plastics, dyes and in chemical synthesis. It serves as a curing agent for epoxy-resins. As a tridentate ligand, it form complexes such as Co(dien)(NO2)3 in coordination chemistry. It is also used as a fuel additive. It reacts with fatty acid resulting in the formation of amidoamines, which are used as corrosion inhibitors in the petroleum industry. It is associated with unsymmetrical dimethylhydrazine to prepare hydyne, which is a propellant for liquid-fuel rockets.

Solubility
Miscible with water and ethanol. Immiscible with ether.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and copper.
RUO – Research Use Only

General References:

  1. Galhoum, A. A.; Mahfouz, M. G.; Rehem, S. T. A.; Gomaa, N. A.; Atia, A. A.; Vincent, T.; Guibal, E. Diethylenetriamine-functionalized chitosan magnetic nano-based particles for the sorption of rare earth metal ions [Nd(III), Dy(III) and Yb(III)]. Cellulose 2015, 22 (4), 2589-2605.
  2. Koizumi, Y.; Hirooka, M.; Ochi, H.; Tokumoto, Y.; Takechi, M.; Hiraoka, A.; Ikeda, Y.; Kumagi, T.; Matsuura, B. Characterization of the biliary tract by virtual ultrasonography constructed by gadolinium ethoxybenzyl diethylenetriamine pentaacetic acid-enhanced magnetic resonance imaging. J Med Ultrason 2015, 42 (2), 185-193.