Product Image
Invitrogen™
Alexa Fluor™ 488 Signal-Amplification Kit for Mouse Antibodies
For sensitive detection of mouse primary antibodies, try our Alexa Fluor® 488 Signal-Amplification Kit, which contains Alexa Fluor 488 rabbitRead more
Have Questions?
Catalog NumberQuantity
A110541 kit
Catalog number A11054
Price (BRL)
2.604,64
Each
Quantity:
1 kit
Price (BRL)
2.604,64
Each
For sensitive detection of mouse primary antibodies, try our Alexa Fluor® 488 Signal-Amplification Kit, which contains Alexa Fluor 488 rabbit anti—mouse IgG and Alexa Fluor 488 goat anti—rabbit IgG conjugates. This kit takes advantage of the intense, photostable fluorescence of Alexa Fluor 488 antibody conjugates and can be used to amplify the signals derived from mouse-derived antibodies. The kit contains a detailed protocol and sufficient reagents for ∼60 assays by flow cytometry or imaging. For added convenience, the kit is designed for use with filters and settings appropriate for fluorescein.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeAlexa Fluor™ 488
Product LineAlexa Fluor
Quantity1 kit
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C) and protect from light.
Citations & References (2)
Citations & References
Abstract
Apical localization of wingless transcripts is required for wingless signaling.
Authors:Simmonds AJ, dosSantos G, Livne-Bar I, Krause HM
Journal:Cell
PubMed ID:11336670
'Many developing and adult tissues are comprised of polarized epithelia. Proteins that are asymmetrically distributed in these cells are thought to be localized by protein trafficking. Here we show that the distribution and function of the signaling protein Wingless is predetermined by the subcellular localization of its mRNA. High-resolution in ... More
The alpha(IIb)beta(3) integrin and GPIb-V-IX complex identify distinct stages in the maturation of CD34(+) cord blood cells to megakaryocytes.
Authors:Lepage A, Leboeuf M, Cazenave JP, de la Salle C, Lanza F, Uzan G
Journal:Blood
PubMed ID:11110688
Megakaryocytopoiesis is a complex multistep process involving cell division, endoreplication, and maturation and resulting in the release of platelets into the blood circulation. Megakaryocytes (MK) progressively express lineage-restricted proteins, some of which play essential roles in platelet physiology. Glycoprotein (GP)Ib-V-IX (CD42) and GPIIb (CD41) are examples of MK-specific proteins having ... More